3-Chloroaniline ≥99%

Danger

C22407-500G C22407-5G C22407-100G
220114-240EA 250.92 USD
220114-240 220114-241 220114-239
3-Chloroaniline ≥99%
3-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 230.5 °C (1013 hPa)
Melting Pt: –10.4 °C
Density: 1.216 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00007765
CAS Number: 108-42-9
EINECS: 203-581-0
UN: 2019
ADR: 6.1,II
Merck Index: 13,02136

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR