(1S)-(+)-Camphor-10-sulfonic acid

Synonyms: ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (1S)-(+)-2-Oxobornane-10-sulphonic acid, D(+)-Camphor-10-sulphonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulphonic acid, (1S)-(+)-10-Camphorsulphonic acid, D-Camphorsulphonic acid

C2107-100G C2107-5G C2107-500G
220114-097EA 88.31 USD
220114-097 220114-099 220114-098
(1S)-(+)-Camphor-10-sulfonic acid
(1S)-(+)-Camphor-10-sulphonic acid
Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 195 °C
MDL Number: MFCD00064157
CAS Number: 3144-16-9
EINECS: 221-554-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR