4'-Chloroacetophenone ≥97%

Warning

Synonyms: p-Chloroacetophenone

C19708-5G C19708-100G C19708-500G
220114-069EA 37.69 USD
220114-069 220114-067 220114-068
4'-Chloroacetophenone ≥97%
4'-Chloroacetophenone
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 18…20 °C
Density: 1.1952 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000624
CAS Number: 99-91-2
EINECS: 202-800-7
UN: 3416
ADR: 6.1,II
Merck Index: 13,02133

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR