ω-Chloroacetophenone ≥98%

Synonyms: 2-Chloroacetophenone, α-Chloroacetophenone, Phenacyl chloride

C19686-100G C19686-500G
220114-063EA 95.95 USD
220114-063 220114-064
ω-Chloroacetophenone ≥98%
ω-Chloroacetophenone
Formula: (C₆H₅)C(O)CH₂Cl
MW: 154.6 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 56.5 °C
Density: 1.324 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000933
CAS Number: 532-27-4
EINECS: 208-531-1
UN: 1697
ADR: 6.1,II
Merck Index: 13,02132

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR