Capric acid ≥98.0%

Warning

Synonyms: Decylic acid, Decanoic acid

C1875-500G C1875-100G
220114-031EA 174.77 USD
220114-031 220114-030
Capric acid ≥98.0%
Capric acid
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 29 - 32 °C
Density: 0.893 g/cm³ (20 °C)
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD00004441
CAS Number: 334-48-5
EINECS: 206-376-4
Merck Index: 13,01764

Order Now

Specification Test Results

Assay 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR