Cyclopropyl methyl ketone ≥99%

Danger

Synonyms: 1-Cyclopropylethanone

C120006-100G C120006-25G
220113-793EA 143.23 USD
220113-793 220113-794
Cyclopropyl methyl ketone ≥99%
Cyclopropyl methyl ketone
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: -68 °C
Density: 0.899 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001297
CAS Number: 765-43-5
EINECS: 212-146-4
UN: 1992
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR