1-Cyclopropylethanol ≥99%

Synonyms: Cyclopropylmethylcarbinol

C119903-5G C119903-25G
220113-792EA 54.23 USD
220113-792 220113-791
1-Cyclopropylethanol ≥99%
1-Cyclopropylethanol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –31.4 °C
Density: 0.883 g/cm³ (20 °C)
MDL Number: MFCD00001300
CAS Number: 765-42-4
EINECS: 212-145-9
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR