4-Sulfamoylbenzoic acid ≥97%

Synonyms: 4-Sulphamoylbenzoic acid

C11804-100G C11804-5G
220113-778EA 216.46 USD
220113-778 220113-779
4-Sulfamoylbenzoic acid ≥97%
4-Sulphamoylbenzoic acid
Formula: C₇H₇NO₄S
MW: 201.2 g/mol
MDL Number: MFCD00007938
CAS Number: 138-41-0
EINECS: 205-327-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR