Bromocyclopropane ≥99%

Synonyms: Cyclopropyl bromide

C117307-25G C117307-5G
220113-772EA 175.54 USD
220113-772 220113-773
Bromocyclopropane ≥99%
Bromocyclopropane
Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 67 - 69 °C (1013 hPa)
Density: 1.505 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001271
CAS Number: 4333-56-6
EINECS: 224-375-7
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR