Cyclopropanecarbonyl chloride ≥98%

Synonyms: Cyclopropanecarboxylic chloride, Cyclopropanecarboxylic acid chloride

C116807-100G C116807-5G C116807-25G
220113-766EA 118.46 USD
220113-766 220113-768 220113-767
Cyclopropanecarbonyl chloride ≥98%
Cyclopropanecarbonyl chloride
Formula: C₄H₅ClO
MW: 104.54 g/mol
Boiling Pt: 119 °C (1013 hPa)
Density: 1.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001277
CAS Number: 4023-34-1
EINECS: 223-684-4
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR