Cyclopropanecarboxylic acid ≥95%

Danger

C116602-25G C116602-100G
220113-762EA 48.89 USD
220113-762 220113-761
Cyclopropanecarboxylic acid ≥95%
Cyclopropanecarboxylic acid
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: 17…19 °C
Density: 1.087 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001287
CAS Number: 1759-53-1
EINECS: 217-162-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR