3-Cyclopentylpropionic acid

Danger

Synonyms: 3-Cyclopentylpropanoic acid, 3-Cylcopentylpropanoic acid

C115908-100ML C115908-25ML
220113-757EA 208.92 USD
220113-757 220113-758
3-Cyclopentylpropionic acid
3-Cyclopentylpropionic acid
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 130 °C (1013 hPa)
Density: 1.01 g/cm³ (17 °C)
MDL Number: MFCD00001392
CAS Number: 140-77-2
EINECS: 205-433-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR