2-Cyclopenten-1-one ≥98%

Synonyms: 2-Cyclopentenone, Cyclopenten-2-one, Cyclopent-2-enone

C112909-25G C112909-5G C112909-100G
220113-057EA 201.38 USD
220113-057 220113-058 220113-056
2-Cyclopenten-1-one ≥98%
2-Cyclopenten-1-one
Formula: C₅H₆O
MW: 82.1 g/mol
Boiling Pt: 64 °C (1013 hPa)
Density: 0.989 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001401
CAS Number: 930-30-3
EINECS: 213-213-0
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR