Cholic acid

Synonyms: 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid

C1129-100G C1129-25G C1129-500G C1129-1KG
220113-052EA 219.45 USD
220113-052 220113-054 220113-055 220113-053
Cholic acid
Cholic acid
Formula: C₂₄H₄₀O₅
MW: 408.58 g/mol
Boiling Pt: 545 °C (1013 hPa)
Melting Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00003672
CAS Number: 81-25-4
EINECS: 201-337-8
Merck Index: 13,02223

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR