Cyclopentanecarboxylic acid ≥99%

Warning

C112003-5G C112003-100G C112003-25G
220113-708EA 43.08 USD
220113-708 220113-706 220113-707
Cyclopentanecarboxylic acid ≥99%
Cyclopentanecarboxylic acid
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 216 °C (1013 hPa)
Melting Pt: 4 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001371
CAS Number: 3400-45-1
EINECS: 222-269-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR