2-Cyclohexen-1-one ≥95%

C102814-10ML C102814-25ML C102814-100ML
220113-574EA 34.68 USD
220113-574 220113-575 220113-573
2-Cyclohexen-1-one ≥95%
2-Cyclohexen-1-one
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 168 °C (1013 hPa)
Density: 0.99 g/cm³ (20 °C)
MDL Number: MFCD00001577
CAS Number: 930-68-7
EINECS: 213-223-5
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR