2-Cyclohexen-1-ol ≥95%

Synonyms: Cyclohex-2-ene-1-ol, Cyclohex-2-enol

C102652-5G C102652-10G C102652-1G
220113-572EA 225.08 USD
220113-572 220113-570 220113-571
2-Cyclohexen-1-ol ≥95%
2-Cyclohexen-1-ol
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 164 - 165 °C (1013 hPa)
Density: 0.9865 g/cm³ (23 °C)
MDL Number: MFCD00001570
CAS Number: 822-67-3
EINECS: 212-501-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR