Cyclohexanone oxime ≥97%

Warning

C102202-100G C102202-500G
220113-562EA 47.06 USD
220113-562 220113-563
Cyclohexanone oxime ≥97%
Cyclohexanone oxime
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 206 °C (1013 hPa)
Melting Pt: 88 - 90 °C
Density: 1.13 g/cm³ (20 °C)
MDL Number: MFCD00001660
CAS Number: 100-64-1
EINECS: 202-874-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR