1,4-Cyclohexanediol ≥99%
Synonyms:
Cyclohexane-1,4-diol
C101206-25G
C101206-100G
220113-537EA
47.38
USD
220113-537
220113-536
1,4-Cyclohexanediol ≥99%
1,4-Cyclohexanediol
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 252 °C (1013 hPa) Density: 1.17 g/cm³ (21.85 °C) |
MDL Number:
MFCD00001448 CAS Number: 556-48-9 EINECS: 209-126-2 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...