Calcium acetate xhydrate ≥99%, ReagentPlus®

Synonyms: Acetic acid calcium salt hydrate, Calcium di(acetate) hydrate, Calcium acetate hydrate

C1000-500G C1000-100G C1000-2.5KG
220113-518EA 171.55 USD
220113-518 220113-516 220113-517
Calcium acetate xhydrate ≥99%, ReagentPlus®
Calcium acetate xhydrate
Formula: (H₃CCOO)₂Ca·nH₂O
MW: 176.18 g/mol
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00150019
CAS Number: 114460-21-8
EINECS: 200-540-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR