Chloramphenicol ≥98%

Danger

Synonyms: Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol

C0378-500G C0378-25G C0378-1KG C0378-100G C0378-5G
220112-651EA 1150.77 USD
220112-651 220112-650 220112-649 220112-648 220112-652
Chloramphenicol ≥98%
Chloramphenicol
Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 150 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078159
CAS Number: 56-75-7
EINECS: 200-287-4
Merck Index: 13,02087

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR