Chloroacetamide

Danger

Synonyms: 2-Chloroacetamide

C0267-1KG C0267-100G C0267-500G
220113-266EA 95.42 USD
220113-266 220113-265 220113-267
Chloroacetamide
Chloroacetamide
Formula: C₂H₄ClNO
MW: 93.51 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 115 - 117 °C
Density: 1.6 g/cm³ (20 °C)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00008027
CAS Number: 79-07-2
EINECS: 201-174-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,02127

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR