2-tert-Butylphenol ≥99%

B99405-50ML B99405-500ML
220112-833EA 28.54 USD
220112-833 220112-832
2-tert-Butylphenol ≥99%
2-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: –7 °C
Density: 0.9823 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002223
CAS Number: 88-18-6
EINECS: 201-807-2
UN: 3145
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR