3,3',4,4'-Benzophenonetetracarboxylic dianhydride

Synonyms: Benzophenone-3,3',4,4'-tetracarboxylic dianhydride

B9750-100G B9750-500G
220112-606EA 62.63 USD
220112-606 220112-607
3,3',4,4'-Benzophenonetetracarboxylic dianhydride
3,3',4,4'-Benzophenonetetracarboxylic dianhydride
Formula: C₁₇H₆O₇
MW: 322.23 g/mol
MDL Number: MFCD00005923
CAS Number: 2421-28-5
EINECS: 219-348-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR