Benzophenone hydrazone ≥96%

B9602-500G B9602-100G
220112-150EA 106.18 USD
220112-150 220112-149
Benzophenone hydrazone ≥96%
Benzophenone hydrazone
Formula: C₁₃H₁₂N₂
MW: 196.25 g/mol
MDL Number: MFCD00007624
CAS Number: 5350-57-2
EINECS: 226-321-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR