Dibutyl disulfide ≥97%

Warning

Synonyms: Dibutyldisulphide, n-Butyl disulphide

B93989-25ML B93989-100ML
220112-564EA 97.78 USD
220112-564 220112-563
Dibutyl disulfide ≥97%
Dibutyl disulphide
Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: -94 °C
Density: 0.9371 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009467
CAS Number: 629-45-8
EINECS: 211-091-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR