Benzo[ghi]perylene

Warning

B9009-25MG-A B9009-100MG-A
220112-767EA 205.31 USD
220112-767 220112-766
Benzo[ghi]perylene
Benzo[ghi]perylene
Formula: C₂₂H₁₂
MW: 276.34 g/mol
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 278…280 °C
MDL Number: MFCD00004135
CAS Number: 191-24-2
EINECS: 205-883-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR