(±)-sec-Butylamine ≥99%

Danger

Synonyms: (±)-2-Butylamine, (±)-2-Aminobutane

B89000-500ML B89000-100ML
220112-748EA 63.65 USD
220112-748 220112-747
(±)-sec-Butylamine ≥99%
(±)-sec-Butylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 63.5 °C (1013 hPa)
Melting Pt: –104 °C
Density: 0.724 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008094
CAS Number: 13952-84-6
EINECS: 237-732-7
UN: 2733
ADR: 3,II
Merck Index: 12,01579

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR