Benzethonium chloride

Danger

Synonyms: Hyamine

B8879-250G B8879-500G B8879-100G
220112-734EA 176.58 USD
220112-734 220112-735 220112-733
Benzethonium chloride
Benzethonium chloride
Formula: C₂₇H₄₂ClNO₂
MW: 448.09 g/mol
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00011742
CAS Number: 121-54-0
EINECS: 204-479-9
UN: 3077
ADR: 9,III
Merck Index: 13,01074

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR