tert-Butyl acetate ≥99%

Danger

Synonyms: 1,1-Dimethylethyl acetate

B88209-100ML B88209-500ML B88209-5ML
220112-727EA 80.23 USD
220112-727 220112-728 220112-729
tert-Butyl acetate ≥99%
tert-Butyl acetate
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 98 °C (1013 hPa)
Melting Pt: 90 - 97 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00008807
CAS Number: 540-88-5
EINECS: 208-760-7
UN: 1123
ADR: 3,II
Merck Index: 13,01536

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR