(±)-3-Buten-2-ol ≥97%

Danger

Synonyms: (±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol

B86400-25G B86400-5G
220112-680EA 177.69 USD
220112-680 220112-681
(±)-3-Buten-2-ol ≥97%
(±)-3-Buten-2-ol
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 96 °C (1013 hPa)
Melting Pt: –100 °C
Density: 0.913 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00004543
CAS Number: 598-32-3
EINECS: 209-929-8
UN: 1986
ADR: 3,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR