Bromothymol blue sodium salt

Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate, Bromothymol blue water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolate

B8630-10G B8630-25G
220112-677EA 76.02 USD
220112-677 220112-678
Bromothymol blue sodium salt
Bromothymol blue sodium salt
Formula: C₂₇H₂₇Br₂NaO₅S
MW: 646.37 g/mol
Melting Pt: ? 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e8e84969735abfa426b8b1c495a534ed9f84651&vid=3491cf8ee21037f844d789eb5cc6be6e7fbbf02b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR