cis-2-Butene-1,4-diol ≥97%

Warning

Synonyms: (Z)-But-2-ene-1,4-diol, (Z)-2-Butene-1,4-diol

B86206-25G B86206-500G
220112-675EA 17.88 USD
220112-675 220112-676
cis-2-Butene-1,4-diol ≥97%
cis-2-Butene-1,4-diol
Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 235 °C (1013 hPa)
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002924
CAS Number: 6117-80-2
EINECS: 228-085-1
Merck Index: 12,02390

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR