2,3-Butanediol ≥98%

Synonyms: 2,3-Dihydroxy butane, (±)-2,3-Butanediol, 2,3-Butylene glykol

B84904-100G B84904-500G
220111-532EA 200.31 USD
220111-532 220111-533
2,3-Butanediol ≥98%
2,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 184 °C (1013 hPa)
Melting Pt: 18…20 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004523
CAS Number: 513-85-9
EINECS: 208-173-6
Merck Index: 12,01602

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR