α-Bromo-m-xylene ≥96%

Synonyms: 3-Methylbenzyl bromide

B83509-25G B83509-5G
220112-519EA 55.14 USD
220112-519 220112-520
α-Bromo-m-xylene ≥96%
α-Bromo-m-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 18 °C
Density: 1.3711 g/cm³ (23 °C)
MDL Number: MFCD00000177
CAS Number: 620-13-3
EINECS: 210-625-2
UN: 1701
ADR: 6.1,II
Merck Index: 12,10223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR