4-Bromoveratrole ≥98%

Synonyms: 1-Bromo-3,4-dimethoxybenzene, 4-Bromo-1,2-dimethoxybenzene

B83355-100G B83355-25G
220112-516EA 70 USD
220112-516 220112-517
4-Bromoveratrole ≥98%
4-Bromoveratrole
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 255 °C (1013 hPa)
Density: 1.5068 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008381
CAS Number: 2859-78-1
EINECS: 220-677-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR