4-Bromoresorcinol ≥97%

Synonyms: 4-Bromobenzene-1,3-diol

B80607-25G B80607-5G
220112-461EA 594.46 USD
220112-461 220112-462
4-Bromoresorcinol ≥97%
4-Bromoresorcinol
Formula: C₆H₅BrO₂
MW: 189.01 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 98 °C
MDL Number: MFCD00002272
CAS Number: 6626-15-9
EINECS: 229-586-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR