4-Bromopyridinium chloride

Warning

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

B80429-1G B80429-5G
220112-458EA 41.03 USD
220112-458 220112-459
4-Bromopyridinium chloride
4-Bromopyridinium chloride
Formula: C₅H₅BrClN
MW: 194.46 g/mol
Melting Pt: 235…238 °C
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR