N-(3-Bromopropyl)phthalimide

B80003-25G B80003-100G
220111-974EA 49.86 USD
220111-974 220111-973
N-(3-Bromopropyl)phthalimide
N-(3-Bromopropyl)phthalimide
Formula: C₁₁H₁₀BrNO₂
MW: 268.11 g/mol
Boiling Pt: 433 °C (1013 hPa)
Melting Pt: 70 - 73 °C
MDL Number: MFCD00005904
CAS Number: 5460-29-7
EINECS: 226-738-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR