3-Brompropylamine hydrobromide

Warning

B79803-25G B79803-100G
220111-972EA 55.03 USD
220111-972 220111-971
3-Brompropylamine hydrobromide
3-Brompropylamine hydrobromide
Formula: BrH₂CCH₂CH₂NH₃Br
MW: 218.92 g/mol
Melting Pt: 173…174 °C
MDL Number: MFCD00012912
CAS Number: 5003-71-4
EINECS: 225-675-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR