2-Bromopropene ≥99%

Synonyms: 2-Propenyl bromide, α-Methylvinyl bromide, Isopropenyl bromide

B78254-10G B78254-50G
220112-439EA 121.69 USD
220112-439 220112-440
2-Bromopropene ≥99%
2-Bromopropene
Formula: C₃H₅Br
MW: 120.98 g/mol
Boiling Pt: 48 °C (1013 hPa)
Melting Pt: –125 °C
Density: 1.39 g/cm³ (9 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000140
CAS Number: 557-93-7
EINECS: 209-185-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR