2-Bromopropane ≥99%

Danger

B78114-500G B78114-100G
220112-436EA 94.77 USD
220112-436 220112-435
2-Bromopropane ≥99%
2-Bromopropane
Formula: H₃CBrCHCH₃
MW: 122.99 g/mol
Boiling Pt: 59 °C (1013 hPa)
Melting Pt: -89 °C
Density: 1.31 g/cm³ (20 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000147
CAS Number: 75-26-3
EINECS: 200-855-1
UN: 2344
ADR: 3,II
Merck Index: 13,05230

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR