1-Bromopropane ≥99%

Danger

Synonyms: Solvent 38, n-Propyl bromide

B78106-5ML B78106-500ML B78106-100ML
220112-432EA 34.57 USD
220112-432 220112-431 220112-430
1-Bromopropane ≥99%
1-Bromopropane
Formula: BrH₂CCH₂CH₃
MW: 122.99 g/mol
Boiling Pt: 71 °C (1013 hPa)
Melting Pt: –108 °C
Density: 1.354 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00000254
CAS Number: 106-94-5
EINECS: 203-445-0
UN: 2344
ADR: 3,II
Merck Index: 12,08030
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63e928c183f1c573c1a2eba4ccb7f1147af103ed&vid=6b2b626154a7592a42b400fdf8ba1c1028a60850&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR