(2-Bromopropyl)benzene ≥96%

Warning

Synonyms: 2-Bromo-1-phenylpropane, (2-Bromopropyl)benzene

B77401-25G B77401-5G
220112-420EA 891.69 USD
220112-420 220112-421
(2-Bromopropyl)benzene ≥96%
(2-Bromopropyl)benzene
Formula: C₆H₅CH₂CHBrCH₃
MW: 199.09 g/mol
Boiling Pt: 107 °C (1013 hPa)
Density: 1.2908 g/cm³ (16 °C)
MDL Number: MFCD00000149
CAS Number: 2114-39-8
EINECS: 218-315-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR