(2-Bromoethoxy)benzene ≥98%

Warning

Synonyms: β-Bromophenetole, 2-Phenoxyethyl bromide, 2-Bromoethyl phenyl ether, ω-Bromophenetole

B75506-100G B75506-25G
220112-385EA 343.54 USD
220112-385 220112-386
(2-Bromoethoxy)benzene ≥98%
(2-Bromoethoxy)benzene
Formula: BrCH₂CH₂OC₆H₅
MW: 201.06 g/mol
Boiling Pt: 144 °C (53 hPa)
Melting Pt: 28…32 °C
Density: 1.3555 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000234
CAS Number: 589-10-6
EINECS: 209-634-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR