6-Bromo-2-naphthol

Synonyms: 6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene

B73406-100G B73406-25G
220112-287EA 138.92 USD
220112-287 220112-288
6-Bromo-2-naphthol
6-Bromo-2-naphthol
Formula: C₁₀H₇BrO
MW: 223.07 g/mol
Boiling Pt: 200…205 °C (20 mmHg)
Melting Pt: 128 °C
MDL Number: MFCD00004081
CAS Number: 15231-91-1
EINECS: 239-279-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR