1-Bromo-4-methylnaphthalene

B72604-5G B72604-25G
220112-262EA 59.34 USD
220112-262 220112-261
1-Bromo-4-methylnaphthalene
1-Bromo-4-methylnaphthalene
Formula: C₁₁H₉Br
MW: 221.1 g/mol
MDL Number: MFCD00003872
CAS Number: 6627-78-7
EINECS: 229-599-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR