2-Bromomesitylene ≥98%

Synonyms: 2,4,6-Trimethylbromobenzene, 2-Bromo-1,3,5-trimethylbenzene, Mesityl bromide, 2-Bromomesitylene

B71608-25G B71608-100G
220112-250EA 45.23 USD
220112-250 220112-249
2-Bromomesitylene ≥98%
2-Bromomesitylene
Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000073
CAS Number: 576-83-0
EINECS: 209-405-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR