4-Bromoisoquinoline ≥98%

Warning

B70202-25G B70202-10G
220112-230EA 168 USD
220112-230 220112-229
4-Bromoisoquinoline ≥98%
4-Bromoisoquinoline
Formula: C₉H₆BrN
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 42 - 42.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006904
CAS Number: 1532-97-4
EINECS: 216-244-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR