1-Bromoheptane ≥99%

Synonyms: 1-Heptyl bromide

B67570-100G B67570-500G
220112-111EA 59.23 USD
220112-111 220112-112
1-Bromoheptane ≥99%
1-Bromoheptane
Formula: BrH₂C(CH₂)₅CH₃
MW: 179.1 g/mol
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: –58 °C
Density: 1.1602 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000273
CAS Number: 629-04-9
EINECS: 211-068-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR